Mivebresib ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: N-(4-(2,4-difluorophenoxy)-3-(6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridin-4-yl)phenyl)ethanesulphonamide,  ABBV-075

A174254-10MG A174254-25MG A174254-50MG A174254-5MG
ALA174254-10MGEA 186.11 USD
ALA174254-10MG ALA174254-25MG ALA174254-50MG ALA174254-5MG
Mivebresib ≥98%, Moligand™
Mivebresib
Formula: C₂₂H₁₉F₂N₃O₄S
MW: 459.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30377200
CAS Number: 1445993-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10389e588fff00ac131197fe6ee1d9f67228741e&vid=10389e588fff00ac131197fe6ee1d9f67228741e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR