2-Amino-4-bromo-3-fluorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Bromo-6-carboxy-2-fluoroaniline, 2-amino-4-bromo-3-fluorobenzoic acid, 4-Bromo-3-fluoroanthranilic acid

A174048-1G A174048-250MG A174048-25G A174048-5G
ALA174048-1GEA 32.21 USD
ALA174048-1G ALA174048250MG ALA174048-25G ALA174048-5G
2-Amino-4-bromo-3-fluorobenzoic acid ≥97%
2-Amino-4-bromo-3-fluorobenzoic acid
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD27664889
CAS Number: 1416013-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee401fe1ba769951d2e3764e5883f2969b76e355&vid=ee401fe1ba769951d2e3764e5883f2969b76e355&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR