(2-Aminopyrimidin-5-yl)methanol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (2-aminopyrimidin-5-yl)methanol

A172565-250MG A172565-100MG A172565-1G
ALA172565250MGEA 1175.06 USD
ALA172565250MG ALA172565100MG ALA172565-1G
(2-Aminopyrimidin-5-yl)methanol ≥97%
(2-Aminopyrimidin-5-yl)methanol
Formula: C₅H₇N₃O
MW: 125.129 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991890
CAS Number: 120747-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db47cb2f181fb32711f47a02b68bb1d1ed014431&vid=db47cb2f181fb32711f47a02b68bb1d1ed014431&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR