RVX-208 ≥97%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Apabetalone, 2-(4-(2-Hydroxyethoxy)-3,5-dimethylphenyl)-5,7-dimethoxyquinazolin-4(3H)-one

A171804-100MG A171804-1MG A171804-250MG A171804-25MG A171804-50MG A171804-5MG A171804-10MG A171804-1G
ALA171804100MGEA 490.11 USD
ALA171804100MG ALA171804-1MG ALA171804250MG ALA171804-25MG ALA171804-50MG ALA171804-5MG ALA171804-10MG ALA171804-1G
RVX-208 ≥97%, Moligand™
RVX-208
Formula: C₂₀H₂₂N₂O₅
MW: 370.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18633270
CAS Number: 1044870-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8ac08e1c0ca6221d030b4bd7be3ff91cde50d4d&vid=a8ac08e1c0ca6221d030b4bd7be3ff91cde50d4d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR