6-Azaspiro[2.5]octane hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-azaspiro[2.5]octane hydrochloride

A171780-1G A171780-250MG A171780-25G A171780-5G
ALA171780-1GEA 56.91 USD
ALA171780-1G ALA171780250MG ALA171780-25G ALA171780-5G
6-Azaspiro[2.5]octane hydrochloride ≥97%
6-Azaspiro[2.5]octane hydrochloride
Formula: C₇H₁₃N·ClH
MW: 147.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14581272
CAS Number: 1037834-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f06b92ee13cf4ec53eb7fcb3532496550e2398&vid=b6f06b92ee13cf4ec53eb7fcb3532496550e2398&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR