Azetidin-3-amine dihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Aminoazetidine dihydrochloride

A171689-250MG A171689-25G A171689-5G A171689-1G
ALA171689250MGEA 25.56 USD
ALA171689250MG ALA171689-25G ALA171689-5G ALA171689-1G
Azetidin-3-amine dihydrochloride ≥97%
Azetidin-3-amine dihydrochloride
Formula: C₃H₁₀Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09910173
CAS Number: 102065-89-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3fd9ea716558c1a20d73bb0c4b179fbfe46a776&vid=f3fd9ea716558c1a20d73bb0c4b179fbfe46a776&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR