2,5-Dioxopyrrolidin-1-yl hex-5-ynoate ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Alkyne NHS ester (hexynoic acid NHS ester), Propargyl-C2-NHS ester

A171448-100MG A171448-50MG
ALA171448100MGEA 326.71 USD
ALA171448100MG ALA171448-50MG
2,5-Dioxopyrrolidin-1-yl hex-5-ynoate ≥95%
2,5-Dioxopyrrolidin-1-yl hex-5-ynoate
Formula: C₁₀H₁₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD28898858
CAS Number: 906564-59-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974308b057bed17ffa6948e3c7ea7582f54013a8&vid=974308b057bed17ffa6948e3c7ea7582f54013a8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR