oxetan-3-ylmethanamine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
3-(Aminomethyl)oxetane, Oxetan-3-ylmethanamine
A171234-100MG
A171234-1G
A171234-250MG
ALA171234100MGEA
175.66
USD
ALA171234100MG
ALA171234-1G
ALA171234250MG
oxetan-3-ylmethanamine ≥97%
oxetan-3-ylmethanamine
Formula:
C₄H₉NO MW: 87.122 g/mol Density: 1.014 g/cm³ Storage Temperature: Freezer |
MDL Number:
MFCD12547197 CAS Number: 6246-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...