3-Acetyl-5-chlorothiophene ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(5-Chlorothiophen-3-yl)ethanone

A171083-1G A171083-250MG A171083-5G
ALA171083-1GEA 53.11 USD
ALA171083-1G ALA171083250MG ALA171083-5G
3-Acetyl-5-chlorothiophene ≥97%
3-Acetyl-5-chlorothiophene
Formula: C₆H₅ClOS
MW: 160.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082792
CAS Number: 58119-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d95103505fc8e9f9e59466b67a80f0f98a81d94c&vid=d95103505fc8e9f9e59466b67a80f0f98a81d94c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR