3,4,5-Trichloropyridin-2-amine ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
2-Amino-3,4,5-trichloropyridine
A170988-1G
A170988-250MG
ALA170988-1GEA
95.86
USD
ALA170988-1G
ALA170988250MG
3,4,5-Trichloropyridin-2-amine ≥95%
3,4,5-Trichloropyridin-2-amine
Formula:
C₅H₃Cl₃N₂ MW: 197.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08705655 CAS Number: 55933-91-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...