1,2-Dihydroacenaphthylene-5-carbaldehyde ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1,2-dihydroacenaphthylene-5-carbaldehyde, 5-Acenaphthenecarboxaldehyde

A170852-1G A170852-5G
ALA170852-1GEA 84.46 USD
ALA170852-1G ALA170852-5G
1,2-Dihydroacenaphthylene-5-carbaldehyde ≥95%
1,2-Dihydroacenaphthylene-5-carbaldehyde
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 367.212°C
Density: 1.243 g/cm³
Flash Pt: 243.636°C
Storage Temperature: Ambient
MDL Number: MFCD00805838
CAS Number: 5345-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49847c6ddfe1a6e1dd85244ffb9ae8636c63a154&vid=49847c6ddfe1a6e1dd85244ffb9ae8636c63a154&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR