2-Amino-5-methylnicotinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A170831-100MG A170831-1G A170831-250MG A170831-25MG
ALA170831100MGEA 97.76 USD
ALA170831100MG ALA170831-1G ALA170831250MG ALA170831-25MG
2-Amino-5-methylnicotinic acid ≥98%
2-Amino-5-methylnicotinic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375376
CAS Number: 532440-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32397776c79503b6ddb54670ce8851dd4d436184&vid=32397776c79503b6ddb54670ce8851dd4d436184&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR