2-Amino-4,5-dimethylthiophene-3-carboxamide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A170707-1G A170707-5G
ALA170707-1GEA 54.9 USD
ALA170707-1G ALA170707-5G
2-Amino-4,5-dimethylthiophene-3-carboxamide ≥98%
2-Amino-4,5-dimethylthiophene-3-carboxamide
Formula: C₇H₁₀N₂OS
Storage Temperature: Ambient
MDL Number: MFCD00617251
CAS Number: 51486-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0215803770185a2bd6db271afb92937d5c7574ea&vid=0215803770185a2bd6db271afb92937d5c7574ea&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR