1-(4-Isocyanatophenyl)ethanone ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
Isocyanic acid 4-Acetylphenyl ester, 4-Acetylphenylisocyanate, 4-Acetylphenyl Isocyanate
A170590-1G
A170590-25G
A170590-5G
ALA170590-1GEA
32.21
USD
ALA170590-1G
ALA170590-25G
ALA170590-5G
1-(4-Isocyanatophenyl)ethanone ≥97%
1-(4-Isocyanatophenyl)ethanone
Formula:
C₉H₇NO₂ Boiling Pt: 139 °C Melting Pt: 37 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013879 CAS Number: 49647-20-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...