1-((4-Methoxy-3-methylphenyl)sulfonyl)piperidine ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: AA92593

A170447-10MG A170447-25MG A170447-50MG A170447-5MG
ALA170447-10MGEA 335.26 USD
ALA170447-10MG ALA170447-25MG ALA170447-50MG ALA170447-5MG
1-((4-Methoxy-3-methylphenyl)sulfonyl)piperidine ≥98% (by HPLC)
1-((4-Methoxy-3-methylphenyl)sulphonyl)piperidine

AA92593 is a selective and competitive OPN4 (melanopsin) antagonist。

Formula: C₁₃H₁₉NO₃S
MW: 269.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02227966
CAS Number: 457961-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27b8e4a875cf39a07589a8a809a500027a9c3652&vid=27b8e4a875cf39a07589a8a809a500027a9c3652&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR