2,6-Dinitro-4-(trifluoromethyl)aniline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-3,5-dinitrobenzotrifluoride, 4-Amino-3,5-dinitro-α,α,α-trifluorotoluene

A170403-1G A170403-5G
ALA170403-1GEA 19.86 USD
ALA170403-1G ALA170403-5G
2,6-Dinitro-4-(trifluoromethyl)aniline ≥97%
2,6-Dinitro-4-(trifluoromethyl)aniline
Formula: C₇H₄F₃N₃O₄
MW: 251.12 g/mol
Melting Pt: 144-149 °C
Storage Temperature: Ambient
MDL Number: MFCD00024260
CAS Number: 445-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecf048f9c259911848dfdd2ecfde01f0a74c5444&vid=ecf048f9c259911848dfdd2ecfde01f0a74c5444&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR