C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
(2,3-Dihydrobenzo-1,4-dioxin-2-yl)methylamine,, 2-Aminomethyl-1,4-benzodioxane, (2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)methanamine, 2-(Aminomethyl)-2,3-dihydro-1,4-benzodioxine
A170391-1G
A170391-250MG
ALA170391-1GEA
54.06
USD
ALA170391-1G
ALA170391250MG
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine ≥97%
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Boiling Pt: 105-110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00203985 CAS Number: 4442-59-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...