1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 7-Azaindole-3-carbonitrile, 3-Cyano-1H-pyrrolo[2,3-b]pyridine,3-Cyano-7-azaindole

A170373-5G A170373-1G A170373-250MG A170373-25G
ALA170373-5GEA 438.81 USD
ALA170373-5G ALA170373-1G ALA170373250MG ALA170373-25G
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile
Formula: C₈H₅N₃
Melting Pt: 258-262°C
Storage Temperature: Ambient
MDL Number: MFCD07778359
CAS Number: 4414-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4299ad565ba29bce07f9ec83ca6cc7402aa8dd52&vid=4299ad565ba29bce07f9ec83ca6cc7402aa8dd52&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR