(1-Aminocyclohexyl)methanol ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Amino-1-(hydroxyethyl)cyclohexane, 1-Amino-1-cyclohexanemethanol

A170341-1G A170341-250MG A170341-50MG
ALA170341-1GEA 309.61 USD
ALA170341-1G ALA170341250MG ALA170341-50MG
(1-Aminocyclohexyl)methanol ≥96%
(1-Aminocyclohexyl)methanol
Formula: C₇H₁₅NO
MW: 129.2 g/mol
Melting Pt: 34-38°C
Storage Temperature: Ambient
MDL Number: MFCD12198034
CAS Number: 4313-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcba9ac6fb11c4623c547a6c731e6b39157e10bb&vid=fcba9ac6fb11c4623c547a6c731e6b39157e10bb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR