2-Acetyl-3-bromothiophene ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A170327-5G A170327-1G A170327-25G
ALA170327-5GEA 62.61 USD
ALA170327-5G ALA170327-1G ALA170327-25G
2-Acetyl-3-bromothiophene ≥97%
2-Acetyl-3-bromothiophene
Formula: C₆H₅BrOS
Boiling Pt: 102-105°C
Melting Pt: 38-41°C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00794287
CAS Number: 42877-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc433115296892dbb303f4711ab5bc36da0edca9&vid=cc433115296892dbb303f4711ab5bc36da0edca9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR