4'-Acetoxy-2'-hydroxyacetophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A170268-1G A170268-50MG
ALA170268-1GEA 6216.71 USD
ALA170268-1G ALA170268-50MG
4'-Acetoxy-2'-hydroxyacetophenone ≥98% (by GC)
4'-Acetoxy-2'-hydroxyacetophenone
Formula: C₁₀H₁₀O₄
Boiling Pt: 303 °C
Melting Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00092225
CAS Number: 42059-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8650dfc23b83a42548099815ad317e2872aefb8&vid=a8650dfc23b83a42548099815ad317e2872aefb8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR