2-Amino-5-methylthiophene-3-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Amino-5-methyl-3-thiophenecarboxylic acid

A170263-1G A170263-250MG
ALA170263-1GEA 221.26 USD
ALA170263-1G ALA170263250MG
2-Amino-5-methylthiophene-3-carboxylic acid ≥97%
2-Amino-5-methylthiophene-3-carboxylic acid
Formula: C₆H₇NO₂S
Storage Temperature: Freezer
MDL Number: MFCD05664138
CAS Number: 41940-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c3e530bb1f68c6bbfa4c98af81d916a6eef7868&vid=0c3e530bb1f68c6bbfa4c98af81d916a6eef7868&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR