4-(Aminomethyl)-N,N-diethylaniline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A170169-100MG A170169-1G A170169-250MG
ALA170169100MGEA 39.81 USD
ALA170169100MG ALA170169-1G ALA170169250MG
4-(Aminomethyl)-N,N-diethylaniline ≥97%
4-(Aminomethyl)-N,N-diethylaniline
Formula: C₁₁H₁₈N₂
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03410985
CAS Number: 40336-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b68a3ff132a8c252a947397a76fb765da87a4c8&vid=4b68a3ff132a8c252a947397a76fb765da87a4c8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR