2-Amino-5,6-dimethyl-4-hydroxypyrimidine ≥96%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A170132-1G
A170132-5G
ALA170132-1GEA
44.56
USD
ALA170132-1G
ALA170132-5G
2-Amino-5,6-dimethyl-4-hydroxypyrimidine ≥96%
2-Amino-5,6-dimethyl-4-hydroxypyrimidine
Formula:
C₆H₉N₃O Storage Temperature: Ambient |
MDL Number:
MFCD00023258 CAS Number: 3977-23-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...