5-Amino-2,4-dichlorophenol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A170104-1G A170104-5G
ALA170104-1GEA 45.51 USD
ALA170104-1G ALA170104-5G
5-Amino-2,4-dichlorophenol ≥97%
5-Amino-2,4-dichlorophenol
Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00186187
CAS Number: 39489-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d74d88682c7ff7ada4fe440bb4ba55b3714b1464&vid=d74d88682c7ff7ada4fe440bb4ba55b3714b1464&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR