3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, 3-aminomethylphenylboronic acid, pinacol ester hydrochloride

A170025-1G A170025-250MG
ALA170025-1GEA 127.21 USD
ALA170025-1G ALA170025250MG
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride ≥98%
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride
Formula: C₁₃H₂₁BClNO₂
MW: 269.58 g/mol
Melting Pt: 188.3°C
Storage Temperature: Refrigerator
MDL Number: MFCD03410294
CAS Number: 380430-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc843ee4c072089c54bbaf5a5fc1fcaaeb97c322&vid=dc843ee4c072089c54bbaf5a5fc1fcaaeb97c322&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR