2-(1-Aminocyclohexyl)acetic acid hydrochloride ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A169998-100MG
A169998-1G
A169998-250MG
A169998-5G
ALA169998100MGEA
11.31
USD
ALA169998100MG
ALA169998-1G
ALA169998250MG
ALA169998-5G
2-(1-Aminocyclohexyl)acetic acid hydrochloride ≥95%
2-(1-Aminocyclohexyl)acetic acid hydrochloride
Formula:
C₈H₁₆ClNO₂ MW: 193.6700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06411715 CAS Number: 37631-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...