Acepromazine maleate ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A169896-10MG
A169896-1G
A169896-250MG
A169896-25MG
A169896-50MG
A169896-5G
ALA169896-10MGEA
52.16
USD
ALA169896-10MG
ALA169896-1G
ALA169896250MG
ALA169896-25MG
ALA169896-50MG
ALA169896-5G
Acepromazine maleate ≥98% (by HPLC)
Acepromazine maleate
Acepromazine is a phenothiazine and an analogue of the raw material of antipsychotic drμg chlorpromazine (Cat. No. C8138). Acepromazine is thoμght to block dopamine receptors in the brain, and inhibit ABCG2 transport protein whose overexpression in tumors is associated with drμg resistance.
Formula:
C₂₃H₂₆N₂O₅S MW: 442.53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082473 CAS Number: 3598-37-6 EINECS: 222-748-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...