(4-Aminobutyl)phosphonic acid ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Aminobutylphosphonic acid, (4-Aminobutyl)phosphonic acid

A169868-100MG A169868-1G A169868-250MG A169868-25MG A169868-5G
ALA169868100MGEA 132.91 USD
ALA169868100MG ALA169868-1G ALA169868250MG ALA169868-25MG ALA169868-5G
(4-Aminobutyl)phosphonic acid ≥95%
(4-Aminobutyl)phosphonic acid
Formula: C₄H₁₂NO₃P
MW: 153.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063426
CAS Number: 35622-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17292daabdae0c1fb4fbd0cadc1112473faea79d&vid=17292daabdae0c1fb4fbd0cadc1112473faea79d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR