4'-(Trifluoromethyl)acetanilide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A169776-100G A169776-1G A169776-25G A169776-5G
ALA169776-100GEA 724.76 USD
ALA169776-100G ALA169776-1G ALA169776-25G ALA169776-5G
4'-(Trifluoromethyl)acetanilide ≥98%
4'-(Trifluoromethyl)acetanilide
Formula: C₉H₈F₃NO
Melting Pt: 150-154°C
Storage Temperature: Ambient
MDL Number: MFCD00013562
CAS Number: 349-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50c7fc155d051ebfdfb5b93f15b1c3963464e75&vid=e50c7fc155d051ebfdfb5b93f15b1c3963464e75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR