(1H-Indol-4-ylmethyl)amine ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
4-Aminomethylindole, (1H-Indol-4-yl)methanamine, 4-(Aminomethyl)indole
A169751-1G
A169751-250MG
A169751-5G
ALA169751-1GEA
91.11
USD
ALA169751-1G
ALA169751250MG
ALA169751-5G
(1H-Indol-4-ylmethyl)amine ≥95%
(1H-Indol-4-ylmethyl)amine
Formula:
C₉H₁₀N₂ MW: 146.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04973297 CAS Number: 3468-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...