1H-Indol-6-ylmethylamine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A169750-1G A169750-250MG A169750-500MG
ALA169750-1GEA 123.41 USD
ALA169750-1G ALA169750250MG ALA169750500MG
1H-Indol-6-ylmethylamine ≥97%
1H-Indol-6-ylmethylamine
Formula: C₉H₁₀N₂
Melting Pt: 135-140 °C
Storage Temperature: Ambient
MDL Number: MFCD06213839
CAS Number: 3468-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c761453d2f780ca57e4f892f0b048aa49791c388&vid=c761453d2f780ca57e4f892f0b048aa49791c388&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR