2-Chloro-6-methoxypyridin-3-amine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-Amino-2-chloro-6-methoxypyridine

A169728-1G A169728-250MG
ALA169728-1GEA 17.96 USD
ALA169728-1G ALA169728250MG
2-Chloro-6-methoxypyridin-3-amine ≥97%
2-Chloro-6-methoxypyridin-3-amine
Formula: C₆H₇ClN₂O
MW: 158.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258809
CAS Number: 34392-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f18086a380750f34089bd807da8b5587a35b25&vid=d4f18086a380750f34089bd807da8b5587a35b25&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR