2-Aminophenanthrene 98

Supplier: ALADDIN SCIENTIFIC
Danger

A169676-1G A169676-250MG A169676-50MG
ALA169676-1GEA 950.86 USD
ALA169676-1G ALA169676250MG ALA169676-50MG
2-Aminophenanthrene 98
2-Aminophenanthrene
Formula: C₁₄H₁₁N
Storage Temperature: Ambient
CAS Number: 3366-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12361aa30a13c3bd42c13b8a1cca9a04b05682dd&vid=12361aa30a13c3bd42c13b8a1cca9a04b05682dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR