4-Allyltoluene ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
p-Allyltoluene
A169644-1G
A169644-5G
ALA169644-1GEA
51.21
USD
ALA169644-1G
ALA169644-5G
4-Allyltoluene ≥97%
4-Allyltoluene
Formula:
C₁₀H₁₂ MW: 132.21 g/mol Density: 0.865 g/cm³ Flash Pt: 54 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00048186 CAS Number: 3333-13-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...