4-Bromo-3-methylisoxazol-5-amine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Bromo-3-methylisoxazol-5-amine, 5-Amino-4-bromo-3-methylisoxazole

A169620-100MG A169620-1G A169620-250MG
ALA169620100MGEA 18.91 USD
ALA169620100MG ALA169620-1G ALA169620250MG
4-Bromo-3-methylisoxazol-5-amine ≥97%
4-Bromo-3-methylisoxazol-5-amine
Formula: C₄H₅BrN₂O
MW: 176.99 g/mol
Melting Pt: 66-71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038913
CAS Number: 33084-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=266615c44aa1319d44170c474bb7b935be8d7193&vid=266615c44aa1319d44170c474bb7b935be8d7193&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR