2-Aminobenzenesulfonamide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A169616-1G A169616-25G
ALA169616-1GEA 17.01 USD
ALA169616-1G ALA169616-25G
2-Aminobenzenesulfonamide ≥98%
2-Aminobenzenesulphonamide
Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 155-157°C
Storage Temperature: Ambient
MDL Number: MFCD00007932
CAS Number: 3306-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba38eee819e84c56d4eb6f94107cd0f081420ca9&vid=ba38eee819e84c56d4eb6f94107cd0f081420ca9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR