1-(3-Aminophenyl)pyrrolidin-2-one ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A169538-100MG A169538-1G A169538-250MG A169538-5G
ALA169538100MGEA 34.11 USD
ALA169538100MG ALA169538-1G ALA169538250MG ALA169538-5G
1-(3-Aminophenyl)pyrrolidin-2-one ≥96%
1-(3-Aminophenyl)pyrrolidin-2-one
Formula: C₁₀H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00468694
CAS Number: 31992-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcb4518ca9838e13393c93c8e8c76df4e7c5abf8&vid=dcb4518ca9838e13393c93c8e8c76df4e7c5abf8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR