2-Amino-5-methyl-4-phenylthiazole ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A169445-10G
A169445-1G
ALA169445-10GEA
719.06
USD
ALA169445-10G
ALA169445-1G
2-Amino-5-methyl-4-phenylthiazole ≥97%
2-Amino-5-methyl-4-phenylthiazole
Formula:
C₁₀H₁₀N₂S Melting Pt: 107-112°C Storage Temperature: Ambient |
MDL Number:
MFCD00617257 CAS Number: 30709-67-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...