4-Acetamidobutyric acid 97

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Acetamidobutanoic acid

A169398-100G A169398-10G A169398-1G A169398-250MG A169398-50G A169398-5G
ALA169398-100GEA 1008.81 USD
ALA169398-100G ALA169398-10G ALA169398-1G ALA169398250MG ALA169398-50G ALA169398-5G
4-Acetamidobutyric acid 97
4-Acetamidobutyric acid
Formula: C₆H₁₁NO₃
MW: 145.16
Melting Pt: 129-131°
Storage Temperature: Ambient
MDL Number: MFCD00020552
CAS Number: 3025-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3232b99842bc0e1d4ece4791308811581364ea78&vid=3232b99842bc0e1d4ece4791308811581364ea78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR