4,5,6,7-Tetrahydro-1,3-benzothiazol-2-amine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4,5,6,7-tetrahydrobenzothiazole

A169332-1G A169332-25G A169332-500MG
ALA169332-1GEA 76.86 USD
ALA169332-1G ALA169332-25G ALA169332500MG
4,5,6,7-Tetrahydro-1,3-benzothiazol-2-amine ≥97%
4,5,6,7-Tetrahydro-1,3-benzothiazol-2-amine
Formula: C₇H₁₀N₂S
MW: 154.24 g/mol
Melting Pt: 86-90°C
Storage Temperature: Ambient
MDL Number: MFCD00022851
CAS Number: 2933-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53af613b236bb8172aa0baf9084915fb8c09f3d1&vid=53af613b236bb8172aa0baf9084915fb8c09f3d1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR