2,6-Dichloro-3-nitropyridin-4-amine ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
4-Amino-2,6-dichloro-3-nitropyridine
A169305-25G
A169305-1G
A169305-250MG
ALA169305-25GEA
223.16
USD
ALA169305-25G
ALA169305-1G
ALA169305250MG
2,6-Dichloro-3-nitropyridin-4-amine ≥98%
2,6-Dichloro-3-nitropyridin-4-amine
Formula:
C₅H₃Cl₂N₃O₂ MW: 208.0022 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03265754 CAS Number: 2897-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...