4-Amino-8-bromo-2-methylquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A169284-1G A169284-250MG
ALA169284-1GEA 201.31 USD
ALA169284-1G ALA169284250MG
4-Amino-8-bromo-2-methylquinoline ≥97%
4-Amino-8-bromo-2-methylquinoline
Formula: C₁₀H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD08669717
CAS Number: 288151-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=124cdff368eb8281d18e2e501d1d328401b519a2&vid=124cdff368eb8281d18e2e501d1d328401b519a2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR