2-Amino-5-chlorophenol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A169264-100G A169264-10G A169264-25G A169264-5G
ALA169264-100GEA 316.26 USD
ALA169264-100G ALA169264-10G ALA169264-25G ALA169264-5G
2-Amino-5-chlorophenol ≥97%
2-Amino-5-chlorophenol

2-Amino-5-chlorophenol can be synthesized from 2-chloro-5-nitrophenol via reduction. It can also be obtained from 1-chloro-4-nitrobenzene by using a bacterial strain LW1. 2-Amino-5-chlorophenol participates in the condensation reaction with acetylferrocene to afford ferrocenyl Schiff bases bearing a phenol group.2-Amino-5-chlorophenol may be used to synthesize 2-amino-5-chloromuconic semialdehyde and benzoxazole derivatives.

Formula: C₆H₆ClNO
Melting Pt: 145-153°C
Storage Temperature: Ambient
MDL Number: MFCD02093863
CAS Number: 28443-50-7
EINECS: 249-020-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0bf419f87275096bf901d961948dbe2f07ed201&vid=f0bf419f87275096bf901d961948dbe2f07ed201&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR