2-Aminobenzothiazol-6-ol ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Hydroxybenzothiazol-2-amine, 2-Amino-6-benzothiazolol, 2-Aminobenzothiazol-6-ol, 6-Hydroxy-2-aminobenzothiazole, 2-Amino-6-hydroxy-1,3-benzothiazole, 2-Amino-6-hydroxybenzothiazole

A169137-100G A169137-1G A169137-25G A169137-5G
ALA169137-100GEA 2600.06 USD
ALA169137-100G ALA169137-1G ALA169137-25G ALA169137-5G
2-Aminobenzothiazol-6-ol ≥95%
2-Aminobenzothiazol-6-ol
Formula: C₇H₆N₂OS
MW: 166.2 g/mol
Melting Pt: 243-250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00511763
CAS Number: 26278-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cebcdd8619d558a50cf19936a8156ba3dfdf5b23&vid=cebcdd8619d558a50cf19936a8156ba3dfdf5b23&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR