(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A169101-1G A169101-250MG
ALA169101-1GEA 20.81 USD
ALA169101-1G ALA169101250MG
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥97%
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid
Formula: C₁₂H₁₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00014378
CAS Number: 2596-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c2255d3043dd30fba60d5fb15ebf8cbda036be&vid=96c2255d3043dd30fba60d5fb15ebf8cbda036be&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR