7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Aza-2-methoxy-1-cycloheptene, O-Methylcaprolactim, 7-Methoxy-3,4,5,6-tetrahydro-2H-azepine

A169044-1G A169044-25G A169044-5G
ALA169044-1GEA 9.41 USD
ALA169044-1G ALA169044-25G ALA169044-5G
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98%
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 53 °C
Density: 0.97 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006938
CAS Number: 2525-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d62f31c03d8e9624062e4d0dbbee476da57fa05&vid=6d62f31c03d8e9624062e4d0dbbee476da57fa05&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR