2-Amino-5-methoxybenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC

A168906-1G A168906-250MG A168906-25G A168906-5G
ALA168906-1GEA 50.26 USD
ALA168906-1G ALA168906250MG ALA168906-25G ALA168906-5G
2-Amino-5-methoxybenzonitrile ≥97%
2-Amino-5-methoxybenzonitrile
Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD05149280
CAS Number: 23842-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c14dcc8d2498171c0f1e25c3cc224ddce7854d7&vid=1c14dcc8d2498171c0f1e25c3cc224ddce7854d7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR