2-Amino-3,5-diiodopyridine ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
3,5-Diiodo-2-pyridylamine, 3,5-Diiodo-2-Pyridinamine, (3,5-Diiodo-2-Pyridyl)Amine, 3,5-Diiodopyridin-2-amine, 2-amino-3,5-diiodopyridine
A168881-250MG
A168881-5G
ALA168881250MGEA
30.31
USD
ALA168881250MG
ALA168881-5G
2-Amino-3,5-diiodopyridine ≥95%
2-Amino-3,5-diiodopyridine
Formula:
C₅H₄I₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD02663852 CAS Number: 23597-15-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...