3'-Isocyanatoacetophenone 95

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Acetylphenyl isocyanate

A168827-1G A168827-5G
ALA168827-1GEA 166.16 USD
ALA168827-1G ALA168827-5G
3'-Isocyanatoacetophenone 95
3'-Isocyanatoacetophenone
Formula: C₉H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00013866
CAS Number: 23138-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fc2d26e38748b8c10960b6acb5f0474c004efd9&vid=7fc2d26e38748b8c10960b6acb5f0474c004efd9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR